Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729024_s1
          See other OLFR464 GE Assays ›
          Gene Symbol:
          Olfr464
          Gene Name:
          olfactory receptor 464
          Gene Aliases:
          MOR240-2
          Chromosome Location:
          Chr.11: 87913920 - 87915003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729024_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr464
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146412.2 NP_666524.2 1 345 87
          GenBank mRNA BC120701.1 - 1 346 40129624 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr464

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005520 NM_146412.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005520 BC120701.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163818 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005520 NM_146412.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005520 BC120701.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P405094 NM_146412.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P405094 BC120701.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27996969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27997007 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163818 - -
          Whole Mouse Genome G4122A A_51_P405094 NM_146412.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P405094 BC120701.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135854 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131835 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09692462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09692458 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09434117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09412488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09412483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09412484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27997012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27996984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09412484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09661826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09434117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09692458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09272246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09556712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27997009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27996972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10389673 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389672 - -
          Affymetrix Mouse Clariom S Array PSR1100031974.mm.1 NM_146412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031974.mm.1 NM_146412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031974.mm.1 TC1100003490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031975.mm.1 NM_146412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031975.mm.1 NM_146412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031975.mm.1 TC1100003490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031973.mm.1 NM_146412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031973.mm.1 NM_146412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031973.mm.1 TC1100003490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031976.mm.1 NM_146412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031976.mm.1 NM_146412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031976.mm.1 TC1100003490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100031977.mm.1 NM_146412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031977.mm.1 NM_146412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100031977.mm.1 TC1100003490.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 349368 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 450324 - -
          Illumina mouse-6 v1.0 expression beadchip 450324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590050 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590050 - -
          mouse-6 v1.1 (Illumina) scl39752.1.1_196-S - -
          Illumina Sentrix mouse-6.v1 bead array 450324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39752.1.1_196-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2590050 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39752.1.1_196-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0