Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729172_s1
          See other OLFR994 GE Assays ›
          Gene Symbol:
          Olfr994
          Gene Name:
          olfactory receptor 994
          Gene Aliases:
          MOR203-4
          Chromosome Location:
          Chr.2: 85429883 - 85436294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729172_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr994
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146433.1 NP_666644.1 1 547 141

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr994

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989175 NM_146433.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124625 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989175 NM_146433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285592 NM_146433.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124625 - -
          Whole Mouse Genome G4122A A_51_P285592 NM_146433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118823 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112426 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01547317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01547308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484517 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484516 - -
          Affymetrix Mouse Clariom S Array PSR0200033686.mm.1 NM_146433.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033686.mm.1 NM_146433.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033686.mm.1 TC0200003895.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410270 - -
          Illumina mouse-6 v1.0 expression beadchip 1410270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613253 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613253 - -
          mouse-6 v1.1 (Illumina) scl4360.1.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4360.1.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613253 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline