Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729176_s1
          See other OLFR998 GE Assays ›
          Gene Symbol:
          Olfr998
          Gene Name:
          olfactory receptor 998
          Gene Aliases:
          MOR175-5
          Chromosome Location:
          Chr.2: 85590510 - 85591521 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729176_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr998
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146436.2 NP_666647.2 1 931 97
          GenBank mRNA BC150719.1 - 1 931 9087630 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr998

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959368 BC150719.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959368 NM_146436.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174984 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959368 BC150719.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959368 NM_146436.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P323732 BC150719.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P323732 NM_146436.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21398594 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174984 - -
          Whole Mouse Genome G4122A A_52_P323732 BC150719.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P323732 NM_146436.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02138841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01594706 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02138834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21398589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01945676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01678395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02138835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01199970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01937191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01199971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01794504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02138839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01950396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01937206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21398590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01199968 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473451 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473450 - -
          Affymetrix Mouse Clariom S Array PSR0200009943.mm.1 NM_146436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009943.mm.1 NM_146436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009943.mm.1 TC0200001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009944.mm.1 NM_146436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009944.mm.1 NM_146436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009944.mm.1 TC0200001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009945.mm.1 NM_146436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009945.mm.1 NM_146436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009945.mm.1 TC0200001241.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1410253 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658244 - -
          mouse-6 v1.1 (Illumina) scl20677.1.241_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410253 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20677.1.241_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410253 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658244 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0