Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729293_s1
          See other OLFR1310 GE Assays ›
          Gene Symbol:
          Olfr1310
          Gene Name:
          olfactory receptor 1310
          Gene Aliases:
          MOR245-3
          Chromosome Location:
          Chr.2: 112008246 - 112009184 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729293_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1310
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146449.1 NP_666660.1 1 167 139
          GenBank mRNA BC119359.1 - 1 193 8734108 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1310

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005445 NM_146449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005445 BC119359.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160341 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005445 NM_146449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005445 BC119359.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366953 NM_146449.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P366953 BC119359.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160341 - -
          Whole Mouse Genome G4122A A_51_P366953 NM_146449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P366953 BC119359.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124891 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01984904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02007992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01303613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01450627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02007991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01383982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02101182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01614882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01383983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01614889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02149108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01383988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485805 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485804 - -
          Affymetrix Mouse Clariom S Array PSR0200036243.mm.1 NM_146449.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036243.mm.1 NM_146449.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036243.mm.1 TC0200004338.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036244.mm.1 TC0200004338.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036242.mm.1 TC0200004338.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724923 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724923 - -
          mouse-6 v1.1 (Illumina) scl4190.1.1_196-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4190.1.1_196-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724923 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline