Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729340_s1
          See other OLFR1231 GE Assays ›
          Gene Symbol:
          Olfr1231
          Gene Name:
          olfactory receptor 1231
          Gene Aliases:
          MOR235-2
          Chromosome Location:
          Chr.2: 89302649 - 89303590 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729340_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1231
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146454.2 NP_666665.2 1 878 95
          GenBank mRNA BC104282.1 - 1 874 95
          BC104283.1 - 1 877 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1231

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021946 NM_146454.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021946 BC104282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021946 BC104283.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140132 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021946 NM_146454.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021946 BC104282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021946 BC104283.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506222 NM_146454.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506222 BC104282.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506222 BC104283.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21413344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140132 - -
          Whole Mouse Genome G4122A A_51_P506222 NM_146454.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506222 BC104282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506222 BC104283.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137934 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114278 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01862264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02164684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02164686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01229516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01229478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01229483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484809 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 368274 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226324 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226324 - -
          mouse-6 v1.1 (Illumina) scl19026.1.204_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19026.1.204_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226324 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline