Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729560_s1
          See other OLFR197 GE Assays ›
          Gene Symbol:
          Olfr197
          Gene Name:
          olfactory receptor 197
          Gene Aliases:
          MOR183-3
          Chromosome Location:
          Chr.16: 59185556 - 59186481 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729560_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr197
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146484.1 NP_666695.1 1 234 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr197

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001398 NM_146484.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168721 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001398 NM_146484.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P133333 NM_146484.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794600 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168721 - -
          Whole Mouse Genome G4122A A_52_P133333 NM_146484.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104640 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122599 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12642466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12699458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12930403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12642460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440169 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440168 - -
          Affymetrix Mouse Clariom S Array PSR1600014467.mm.1 NM_146484.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014467.mm.1 NM_146484.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014467.mm.1 TC1600001794.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 773020 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4210672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232267 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232267 - -
          mouse-6 v1.1 (Illumina) scl48357.3.18_31-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48357.3.18_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2956500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2956500 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl48357.3.18_31-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0