Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729763_s1
          See other OLFR95 GE Assays ›
          Gene Symbol:
          Olfr95
          Gene Name:
          olfactory receptor 95
          Gene Aliases:
          MOR263-6
          Chromosome Location:
          Chr.17: 37210913 - 37211851 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729763_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146513.1 NP_666724.1 1 44 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr95

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078810 NM_146513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061555 NM_146513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061555 NM_146513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078810 NM_146513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144740 NM_146513.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167858 - -
          Whole Mouse Genome G4122A A_51_P144740 NM_146513.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107511 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201601 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13107356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13107354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13163084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13418037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13315372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13418036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450865 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450864 - -
          Affymetrix Mouse Clariom S Array PSR1700019547.mm.1 NM_146513.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019547.mm.1 NM_146513.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019547.mm.1 TC1700002059.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 880532 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840458 - -
          Illumina mouse-6 v1.0 expression beadchip 6840458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233741 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233741 - -
          mouse-6 v1.1 (Illumina) scl0258506.1_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258506.1_71-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233741 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233741 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0