Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729797_s1
          See other OLFR523 GE Assays ›
          Gene Symbol:
          Olfr523
          Gene Name:
          olfactory receptor 523
          Gene Aliases:
          MOR104-4
          Chromosome Location:
          Chr.7: 140176104 - 140177057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729797_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr523
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146518.1 NP_666729.1 1 651 79
          GenBank mRNA BC119240.1 - 1 694 8733989 79
          BC120529.1 - 1 694 40129452 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr523

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138457 NM_146518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138457 BC119240.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138457 BC120529.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175752 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138457 NM_146518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138457 BC119240.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138457 BC120529.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P645805 NM_146518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P645805 BC119240.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P645805 BC120529.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582699 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175752 - -
          Whole Mouse Genome G4122A A_52_P645805 NM_146518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645805 BC119240.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645805 BC120529.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133000 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226551 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06221937 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05913578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06623465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06226551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06599373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06129292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06369376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05903433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10558613 - -
          Affymetrix Mouse Gene 1.1 ST Array 10558612 - -
          Affymetrix Mouse Clariom S Array PSR0700017620.mm.1 TC0700002005.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 721827 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7040176 - -
          Illumina mouse-6 v1.0 expression beadchip 7040176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251687 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251687 - -
          mouse-6 v1.1 (Illumina) scl31917.1.1_217-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31917.1.1_217-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline