Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729918_s1
          See other OLFR1368 GE Assays ›
          Gene Symbol:
          Olfr1368
          Gene Name:
          olfactory receptor 1368
          Gene Aliases:
          MOR256-15
          Chromosome Location:
          Chr.13: 21142111 - 21143055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729918_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1368
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146534.1 NP_666745.1 1 457 67
          GenBank mRNA BC125382.1 - 1 483 40129897 67
          BC125384.1 - 1 483 40129899 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1368

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053943 NM_146534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053943 BC125382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053943 BC125384.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163861 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053943 NM_146534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053943 BC125382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053943 BC125384.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460562 NM_146534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460562 BC125382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P460562 BC125384.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28871314 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163861 - -
          Whole Mouse Genome G4122A A_51_P460562 NM_146534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460562 BC125382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460562 BC125384.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130314 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10999143 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10999153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10999132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11067571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11067560 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11067562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11067566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28871287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10865612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10827121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10919836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10451645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10999147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28871274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407998 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407997 - -
          Affymetrix Mouse Clariom S Array PSR1300009705.mm.1 NM_146534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009705.mm.1 NM_146534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009705.mm.1 TC1300001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009704.mm.1 TC1300001531.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009706.mm.1 TC1300001531.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 778964 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030131 - -
          Illumina mouse-6 v1.0 expression beadchip 2030131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710507 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710507 - -
          mouse-6 v1.1 (Illumina) scl44242.1.1_80-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44242.1.1_80-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710507 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0