Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730041_s1
          See other OLFR828 GE Assays ›
          Gene Symbol:
          Olfr828
          Gene Name:
          olfactory receptor 828
          Gene Aliases:
          MOR149-1
          Chromosome Location:
          Chr.9: 18815354 - 18816292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730041_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr828
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146605.1 NP_666816.1 1 843 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr828

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184841 NM_146605.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165386 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184841 NM_146605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233885 NM_146605.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338557 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165386 - -
          Whole Mouse Genome G4122A A_51_P233885 NM_146605.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128741 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165386 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08020949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08020962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07908230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08020956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07908228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07856881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07908227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07856883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07841008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591181 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591180 - -
          Affymetrix Mouse Clariom S Array PSR0900015222.mm.1 NM_146605.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015222.mm.1 NM_146605.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015222.mm.1 TC0900001811.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 523955 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234579 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234579 - -
          mouse-6 v1.1 (Illumina) scl6695.1.1_217-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6695.1.1_217-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234579 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline