Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730071_s1
          See other OLFR984 GE Assays ›
          Gene Symbol:
          Olfr984
          Gene Name:
          olfactory receptor 984
          Gene Aliases:
          MOR239-6
          Chromosome Location:
          Chr.9: 40100544 - 40156029 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730071_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr984
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146608.1 NP_666819.1 1 848 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr984

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159139 NM_146608.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167314 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159139 NM_146608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P659241 NM_146608.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441316 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167314 - -
          Whole Mouse Genome G4122A A_52_P659241 NM_146608.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130826 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124716 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167314 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07701012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07701024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07667646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07631001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07701017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07701010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592468 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592467 - -
          Affymetrix Mouse Clariom S Array PSR0900017276.mm.1 NM_146608.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017276.mm.1 NM_146608.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017276.mm.1 TC0900002092.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 422771 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940019 - -
          Illumina mouse-6 v1.0 expression beadchip 940019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256542 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256542 - -
          mouse-6 v1.1 (Illumina) scl36004.2.191_208-S - -
          Illumina Sentrix mouse-6.v1 bead array 940019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36004.2.191_208-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256542 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline