Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730129_s1
          See other OLFR307 GE Assays ›
          Gene Symbol:
          Olfr307
          Gene Name:
          olfactory receptor 307
          Gene Aliases:
          MOR219-1
          Chromosome Location:
          Chr.7: 86335453 - 86336394 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730129_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr307
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146617.1 NP_666828.1 1 49 77
          GenBank mRNA BC132055.1 - 1 72 40130332 77
          BC132437.1 - 1 72 40130714 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr307

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122110 BC132437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122110 NM_146617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122110 BC132055.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135872 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122110 BC132437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122110 NM_146617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122110 BC132055.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P482023 BC132437.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P482023 NM_146617.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P482023 BC132055.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294299 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135872 - -
          Whole Mouse Genome G4122A A_52_P482023 BC132437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482023 NM_146617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P482023 BC132055.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114117 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06640102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200030 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200034 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06640109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06640110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06052164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06640109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06637745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565390 - -
          Affymetrix Mouse Gene 1.1 ST Array 10565389 - -
          Affymetrix Mouse Clariom S Array PSR0700031230.mm.1 NM_146617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031230.mm.1 NM_146617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031230.mm.1 TC0700003691.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664964 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258690 - -
          mouse-6 v1.1 (Illumina) scl0258610.1_145-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258610.1_145-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258690 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0