Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730151_s1
          See other OLFR303 GE Assays ›
          Gene Symbol:
          Olfr303
          Gene Name:
          olfactory receptor 303
          Gene Aliases:
          MOR104-2
          Chromosome Location:
          Chr.7: 86393766 - 86395529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730151_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr303
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146619.1 NP_666830.1 1 444 99
          GenBank mRNA BC132005.1 - 1 483 40130282 99
          BC145991.1 - 1 483 40131294 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr303

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991431 NM_146619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991431 BC132005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991431 BC145991.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373907 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126052 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991431 NM_146619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991431 BC132005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991431 BC145991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634930 NM_146619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634930 BC132005.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634930 BC145991.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294555 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126052 - -
          Whole Mouse Genome G4122A A_52_P634930 NM_146619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634930 BC132005.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634930 BC145991.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113319 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06372582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06460501 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06460499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06394747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06372578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06372574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294559 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565396 - -
          Affymetrix Mouse Gene 1.1 ST Array 10565395 - -
          Affymetrix Mouse Clariom S Array PSR0700031237.mm.1 TC0700003695.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700031235.mm.1 TC0700003695.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700031236.mm.1 NM_146619.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031236.mm.1 NM_146619.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700031236.mm.1 TC0700003695.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 707928 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4540121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244318 - -
          mouse-6 v1.1 (Illumina) scl31068.1.118_192-S - -
          Illumina Sentrix mouse-6.v1 bead array 4540121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31068.1.118_192-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline