Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730309_s1
          See other OLFR1138 GE Assays ›
          Gene Symbol:
          Olfr1138
          Gene Name:
          olfactory receptor 1138
          Gene Aliases:
          MOR177-8
          Chromosome Location:
          Chr.2: 87737387 - 87738322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730309_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146639.1 NP_666850.1 1 447 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1138

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172182 NM_146639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172182 NM_146639.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P554496 NM_146639.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407083 - -
          Whole Mouse Genome G4122A A_52_P554496 NM_146639.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01363868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484692 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484691 - -
          Affymetrix Mouse Clariom S Array PSR0200033892.mm.1 NM_146639.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033892.mm.1 NM_146639.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033892.mm.1 TC0200003994.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6980465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224920 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224920 - -
          mouse-6 v1.1 (Illumina) scl0258632.1_243-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258632.1_243-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224920 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline