Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730328_s1
          See other OLFR1140 GE Assays ›
          Gene Symbol:
          Olfr1140
          Gene Name:
          olfactory receptor 1140
          Gene Aliases:
          MOR177-6
          Chromosome Location:
          Chr.2: 87746161 - 87747180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730328_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1140
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146642.2 NP_666853.2 1 582 123
          GenBank mRNA BC120608.1 - 1 582 40129531 123
          BC120610.1 - 1 582 40129533 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1140

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129444 NM_146642.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129444 BC120608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2129444 BC120610.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129444 NM_146642.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129444 BC120608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2129444 BC120610.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P71645 NM_146642.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P71645 BC120608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P71645 BC120610.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21407105 - -
          Whole Mouse Genome G4122A A_52_P71645 NM_146642.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P71645 BC120608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P71645 BC120610.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121498 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01181624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01409693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01301861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473561 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473559 - -
          Affymetrix Mouse Clariom S Array PSR0200010072.mm.1 NM_146642.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010072.mm.1 NM_146642.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010072.mm.1 TC0200001300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010071.mm.1 NM_146642.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010071.mm.1 NM_146642.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010071.mm.1 TC0200001300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010070.mm.1 NM_146642.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010070.mm.1 NM_146642.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010070.mm.1 TC0200001300.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769908 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769908 - -
          mouse-6 v1.1 (Illumina) scl0258635.1_115-S - -
          Illumina Sentrix mouse-6.v1 bead array 110270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258635.1_115-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2875611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769908 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2875611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline