Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730522_s1
          See other OLFR736 GE Assays ›
          Gene Symbol:
          Olfr736
          Gene Name:
          olfactory receptor 736
          Gene Aliases:
          MOR106-5
          Chromosome Location:
          Chr.14: 50392758 - 50393696 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730522_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr736
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146666.1 NP_666877.1 1 447 102
          GenBank mRNA BC110467.2 - 1 447 40041632 102
          BC127601.1 - 1 447 40041634 102
          BC129795.1 - 1 445 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr736

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC127601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC129795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 NM_146666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC127601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC129795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 NM_146666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 247632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170216 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119415 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173798 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC127601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC129795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 NM_146666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC110467.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC127601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC129795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 NM_146666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC127601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC129795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 NM_146666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC127601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC129795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 NM_146666.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597283 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161246 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119415 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173798 - -
          Whole Mouse Genome G4122A A_52_P549135 BC127601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 BC129795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 NM_146666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 BC110467.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 BC127601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 BC129795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 NM_146666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549135 BC110467.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132246 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161246 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119415 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11510528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11606095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11510523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11510514 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11510522 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11510518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11606095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11510520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11304518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414474 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414473 - -
          Affymetrix Mouse Clariom S Array PSR1400004418.mm.1 NM_146666.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004418.mm.1 NM_146666.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004418.mm.1 TC1400000553.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428256 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6110204 - -
          Illumina mouse-6 v1.0 expression beadchip 6110204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220803 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220803 - -
          mouse-6 v1.1 (Illumina) scl46376.3.215_44-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46376.3.215_44-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220803 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline