Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730576_s1
          See other OLFR820 GE Assays ›
          Gene Symbol:
          Olfr820
          Gene Name:
          olfactory receptor 820
          Gene Aliases:
          MOR116-1
          Chromosome Location:
          Chr.10: 130017363 - 130018301 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730576_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr820
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146675.1 NP_666886.1 1 469 96
          GenBank mRNA BC119398.1 - 1 531 8734147 96
          BC119400.1 - 1 531 8734149 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr820

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127030 NM_146675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127030 BC119398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127030 BC119400.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335370 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150440 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127030 NM_146675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127030 BC119398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127030 BC119400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P218857 NM_146675.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P218857 BC119398.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P218857 BC119400.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548046 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150440 - -
          Whole Mouse Genome G4122A A_51_P218857 NM_146675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218857 BC119398.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218857 BC119400.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105141 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08878229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08743327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08743331 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08743319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08743322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08537893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08835524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08674487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08407012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08917359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08743331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367527 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367526 - -
          Affymetrix Mouse Clariom S Array PSR1000012058.mm.1 NM_146675.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012058.mm.1 NM_146675.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012058.mm.1 TC1000001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012057.mm.1 TC1000001657.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324855 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225952 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225952 - -
          mouse-6 v1.1 (Illumina) scl5598.1.1_312-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5598.1.1_312-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2823628 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2823628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0