Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730612_s1
          See other OLFR1427 GE Assays ›
          Gene Symbol:
          Olfr1427
          Gene Name:
          olfactory receptor 1427
          Gene Aliases:
          MOR239-4
          Chromosome Location:
          Chr.19: 12098702 - 12099637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730612_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1427
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146679.1 NP_666890.1 1 848 87
          GenBank mRNA BC127962.1 - 1 852 40134881 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1427

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50207 NM_146679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50207 NM_146679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113779 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50207 NM_146679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50207 BC127962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50207 NM_146679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P50207 NM_146679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P50207 NM_146679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959503 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113779 - -
          Whole Mouse Genome G4122A A_52_P50207 NM_146679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50207 BC127962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50207 NM_146679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50207 BC127962.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129869 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109884 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113779 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14295889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14295896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14482904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14295886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14152985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14199843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14187002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14285211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14187001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14482904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466287 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466286 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768980 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768980 - -
          mouse-6 v1.1 (Illumina) scl52712.1.136_161-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52712.1.136_161-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2768980 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline