Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730616_s1
          See other OLFR1423 GE Assays ›
          Gene Symbol:
          Olfr1423
          Gene Name:
          olfactory receptor 1423
          Gene Aliases:
          MOR239-3
          Chromosome Location:
          Chr.19: 12035808 - 12036740 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730616_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1423
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146680.1 NP_666891.1 1 249 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1423

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060682 NM_146680.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167043 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060682 NM_146680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159986 NM_146680.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959219 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167043 - -
          Whole Mouse Genome G4122A A_51_P159986 NM_146680.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114120 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14308585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14308574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14308590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14168892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14275424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14308583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14293566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14330386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14446023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14274125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14494500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14328343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14228507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14327722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14150048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14228499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466277 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466276 - -
          Affymetrix Mouse Clariom S Array PSR1900010762.mm.1 NM_146680.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010762.mm.1 NM_146680.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010762.mm.1 TC1900001150.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 741564 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4560575 - -
          Illumina mouse-6 v1.0 expression beadchip 4560575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246989 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246989 - -
          mouse-6 v1.1 (Illumina) scl52715.1.136_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52715.1.136_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246989 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline