Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00730829_s1
          See other OLFR385 GE Assays ›
          Gene Symbol:
          Olfr385
          Gene Name:
          olfactory receptor 385
          Gene Aliases:
          MOR135-3
          Chromosome Location:
          Chr.11: 73588798 - 73589736 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00730829_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr385
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147023.1 NP_667234.1 1 410 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr385

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115195 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P498178 NM_147023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917566 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115195 - -
          Whole Mouse Genome G4122A A_52_P498178 NM_147023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P498178 NM_147023.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131158 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123326 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724157 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09724171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09500700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09500701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09465044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09465049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09393795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09724157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09393796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917543 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388281 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388280 - -
          Affymetrix Mouse Clariom S Array PSR1100029352.mm.1 NM_147023.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029352.mm.1 NM_147023.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029352.mm.1 TC1100003173.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100510315 - -
          MouseWG-6 v2 Expression BeadChip 6380048 - -
          Illumina mouse-6 v1.0 expression beadchip 6380048 - -
          Illumina mouse-6 v1.0 expression beadchip 100510315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2672020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257775 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2672020 - -
          mouse-6 v1.1 (Illumina) scl39955.1.1_31-S - -
          mouse-6 v1.1 (Illumina) GI_33239299-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380048 - -
          Illumina Sentrix mouse-6.v1 bead array 100510315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39955.1.1_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100510315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2672020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2672020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline