Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731198_s1
          See other 9330132A10RIK GE Assays ›
          Gene Symbol:
          9330132A10Rik
          Gene Name:
          RIKEN cDNA 9330132A10 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731198_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330132A10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK033982.1 - 3562 147
          BC098197.1 - 3288 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630619 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31911390 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111607 - -
          Whole Mouse Genome G4122A A_52_P553421 BC098197.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P553421 AK033982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P553421 AK083197.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132084 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31911304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31911299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13981109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31911302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31911301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31911305 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460124 - -
          Affymetrix Mouse Gene 1.1 ST Array 10460123 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 481119 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3800685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618514 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618514 - -
          mouse-6 v1.1 (Illumina) scl51173.1.1_93-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51173.1.1_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800685 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618514 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline