Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731567_m1
          See other CYP3A11 GE Assays ›
          Gene Symbol:
          Cyp3a11
          Gene Name:
          cytochrome P450, family 3, subfamily a, polypeptide 11
          Gene Aliases:
          AI256190, Cyp3a, IIIAm1, Pcn
          Chromosome Location:
          Chr.5: 145854607 - 145879854 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731567_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp3a11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007818.3 NP_031844.1 10 - 11 1108 128
          GenBank mRNA AK004861.2 - 2058 128
          AK149433.1 - 10 - 11 1119 128
          BC010528.1 - 10 - 11 1108 4194909 128
          BC094062.1 - 10 - 11 1104 4162188 128
          X60452.1 - 10 - 11 1104 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003600.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection P450 Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp3a11

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          demethylation
          monooxygenase activity
          iron ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          heme binding
          demethylase activity
          metal ion binding
          testosterone 6-beta-hydroxylase activity
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121225 AK004861.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121225 NM_007818.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 BC010528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 BC094062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 X60452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 AK004861.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 AK149433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 BC010528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 BC094062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 X60452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 AK004861.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P355301 AK149433.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860778 AK038602.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63062 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126128 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 BC010528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 BC094062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 X60452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 AK004861.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 AK149433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 BC010528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 BC094062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 X60452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 AK004861.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P355301 AK149433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121225 AK004861.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121225 NM_007818.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 BC010528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 BC094062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 X60452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 AK004861.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 AK149433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 BC010528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 BC094062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 X60452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 AK004861.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P355301 AK149433.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24166326 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126128 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05780076 - -
          Whole Mouse Genome G4122A A_51_P355301 BC010528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 BC094062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 X60452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 AK004861.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 AK149433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 BC010528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 BC094062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 X60452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 AK004861.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355301 AK149433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118461 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04423732 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04423734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04423735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04423731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24166310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04429687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04932768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04423734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04264688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04423731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04917134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04765774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04409706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04409703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04639207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04765782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24166320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535709 - -
          Affymetrix HT MG-430 PM Array Plate 1416809_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535704 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535669 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535683 - -
          Mouse Genome 430 2.0 Array 1416809_at - -
          Mouse Genome 430 2.0 Array 1416809_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13112_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13112_at - -
          Affymetrix Mouse Clariom S Array PSR0500033054.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033054.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033054.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033048.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033048.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033048.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033053.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033053.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033053.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033056.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033056.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033056.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033046.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033046.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033046.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033058.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033058.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033058.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033051.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033051.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033051.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033052.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033052.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033052.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033045.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033045.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033045.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033047.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033047.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033047.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033049.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033049.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033049.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033050.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033050.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033050.mm.1 NM_007818.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033059.mm.1 TC0500003600.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033059.mm.1 NM_007818.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033059.mm.1 NM_007818.3 Refseq
          Mouse Expression Set 430 1416809_PM_at - -
          Mouse Expression Set 430 1416809_at - -
          Murine Genome U74v2 Set 101638_s_at - -
          Murine Genome U74v2 Set 93770_PM_at - -
          Murine Genome U74v2 Set 101638_PM_s_at - -
          Murine Genome U74v2 Set 93770_at - -
          Mouse Genome 430A 2.0 Array 1416809_at - -
          Mouse Genome 430A 2.0 Array 1416809_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416809_at - -
          Affymetrix HT Mouse Genome 430A Array 1416809_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753183 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753183 - -
          mouse-6 v1.1 (Illumina) scl0013112.1_193-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013112.1_193-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753183 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0