Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731652_g1
          See other GZMF GE Assays ›
          Gene Symbol:
          Gzmf
          Gene Name:
          granzyme F
          Gene Aliases:
          CCP4, Ctla-7, Ctla7, MCSP-3
          Chromosome Location:
          Chr.14: 56205263 - 56211407 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731652_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gzmf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010374.3 NP_034504.1 2 - 3 255 149
          GenBank mRNA BC100311.1 - 2 - 3 255 30523018 149
          BC145435.1 - 2 - 3 231 9053993 149
          J03257.1 - 2 - 3 265 149
          X12823.1 - 2 - 3 206 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002865.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gzmf

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          immune response
          granzyme-mediated apoptotic signaling pathway
          protein processing
          cytolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 X12823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 BC145435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 J03257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 NM_010374.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 X12823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 BC145435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 J03257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 NM_010374.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105158 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 X12823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 BC145435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 J03257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 NM_010374.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 BC100311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 X12823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 BC145435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 J03257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 NM_010374.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 X12823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 BC145435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 J03257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 NM_010374.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 X12823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 BC145435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 J03257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 NM_010374.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08120499 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105061 - -
          Whole Mouse Genome G4122A A_51_P190961 X12823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 BC145435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 J03257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 NM_010374.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 BC100311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 X12823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 BC145435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 J03257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 NM_010374.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P190961 BC100311.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116474 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08120499 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11460643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11533564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11745705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11745703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11460648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420297 - -
          Affymetrix HT MG-430 PM Array Plate 1418679_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420295 - -
          Mouse Genome 430 2.0 Array 1418679_at - -
          Mouse Genome 430 2.0 Array 1418679_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14943_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14943_at - -
          Affymetrix Mouse Clariom S Array PSR1400016329.mm.1 NM_010374.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016329.mm.1 NM_010374.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016329.mm.1 TC1400002865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016330.mm.1 NM_010374.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016330.mm.1 NM_010374.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016330.mm.1 TC1400002865.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016331.mm.1 NM_010374.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016331.mm.1 NM_010374.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016331.mm.1 TC1400002865.mm.1 MTA TC
          Mouse Expression Set 430 1418679_at - -
          Mouse Expression Set 430 1418679_PM_at - -
          Murine Genome U74v2 Set 102822_PM_at - -
          Murine Genome U74v2 Set 102822_at - -
          Mouse Genome 430A 2.0 Array 1418679_PM_at - -
          Mouse Genome 430A 2.0 Array 1418679_at - -
          Affymetrix HT Mouse Genome 430A Array 1418679_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418679_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 912021 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6200008 - -
          Illumina mouse-6 v1.0 expression beadchip 6200008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629794 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629794 - -
          mouse-6 v1.1 (Illumina) scl45511.2.1_44-S - -
          Illumina Sentrix mouse-6.v1 bead array 6200008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45511.2.1_44-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629794 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629794 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline