Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731842_g1
          See other CES2A GE Assays ›
          Gene Symbol:
          Ces2a
          Gene Name:
          carboxylesterase 2A
          Gene Aliases:
          9130231C15Rik, AI266984, Ces6
          Chromosome Location:
          Chr.8: 104734003 - 104741634 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731842_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190330.1 NP_001177259.1 9 - 10 1390 98
          NM_133960.5 NP_598721.1 10 - 11 1489 98
          GenBank mRNA AK078953.1 - 10 - 11 1485 98
          AK165784.1 - 9 - 10 1380 98
          BC021609.1 - 2 - 3 260 98
          BC024082.1 - 10 - 11 1455 5100146 98
          BC024491.1 - 10 - 11 1464 4981579 98
          BC024517.1 - 10 - 11 1462 4984109 98
          BC025537.1 - 10 - 11 1475 5309880 98
          BC025812.1 - 10 - 11 1462 5096915 98
          BC026643.1 - 10 - 11 1462 4986124 98
          BC031295.1 - 10 - 11 1455 5100144 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001191.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces2a

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2293013 BC031295.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2293013 BC026643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2293013 BC024517.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2293013 BC025537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2293013 BC025812.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2293013 BC031295.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2293013 BC026643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2293013 BC024517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2293013 BC025537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2293013 BC025812.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26129996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26129999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26129994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26130023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06624822 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124967 - -
          Whole Mouse Genome G4122A A_51_P443339 BC031295.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443339 BC026643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443339 BC024517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443339 BC025537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443339 BC025812.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135246 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372434 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372430 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372426 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07372427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26129984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26130001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06818535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06843985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06843983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07098069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07372427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07372420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06931447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07396888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06699850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06931455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07187986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06818540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06795749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06931451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26129990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06818538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26130055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574505 - -
          Affymetrix HT MG-430 PM Array Plate 1449375_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574498 - -
          Mouse Genome 430 2.0 Array 1449375_PM_at - -
          Mouse Genome 430 2.0 Array 1449375_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102022_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102022_at - -
          Affymetrix Mouse Clariom S Array PSR0800009254.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009254.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009254.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009254.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009254.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009256.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009256.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009256.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009256.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009256.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009250.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009250.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009250.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009250.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009250.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009251.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009251.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009251.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009251.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009251.mm.1 TC0800001191.mm.1 MTA TC
          Mouse Expression Set 430 1449375_at - -
          Mouse Expression Set 430 1449375_PM_at - -
          Mouse Genome 430A 2.0 Array 1449375_PM_at - -
          Mouse Genome 430A 2.0 Array 1449375_at - -
          Affymetrix HT Mouse Genome 430A Array 1449375_at - -
          Affymetrix HT Mouse Genome 430A Array 1449375_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 750552 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760152 - -
          Illumina mouse-6 v1.0 expression beadchip 4760152 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259503 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259503 - -
          mouse-6 v1.1 (Illumina) scl33434.12_331-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33434.12_331-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259503 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline