Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00731896_s1
          See other OLFR1058 GE Assays ›
          Gene Symbol:
          Olfr1058
          Gene Name:
          olfactory receptor 1058
          Gene Aliases:
          MOR190-2
          Chromosome Location:
          Chr.2: 86385466 - 86386416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00731896_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1058
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146391.2 NP_666503.2 1 64 151

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1058

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044638 NM_146391.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044638 NM_146391.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116647 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01759382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01410207 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01410215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21402089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01410215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01679604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01168276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21402042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484603 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484602 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484594 - -
          Affymetrix Mouse Clariom S Array PSR0200033760.mm.1 NM_146391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033760.mm.1 NM_146391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033760.mm.1 TC0200003934.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410021 - -
          Illumina mouse-6 v1.0 expression beadchip 1410021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773759 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773759 - -
          mouse-6 v1.1 (Illumina) scl0258386.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258386.1_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2773758 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773758 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline