Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732122_s1
          See other OLFR1209 GE Assays ›
          Gene Symbol:
          Olfr1209
          Gene Name:
          olfactory receptor 1209
          Gene Aliases:
          MOR230-7
          Chromosome Location:
          Chr.2: 88909459 - 88910391 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732122_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1209
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146461.2 NP_666672.2 1 47 105
          GenBank mRNA BC104269.1 - 1 47 40040821 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1209

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 BC104270.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 BC104269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 BC104270.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 BC104269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99699 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 BC104270.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 BC104269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 NM_146461.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 BC104270.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 BC104269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 BC104270.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 BC104269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 BC104270.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 BC104269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411712 - -
          Whole Mouse Genome G4122A A_51_P123475 BC104270.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123475 BC104269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123475 NM_146461.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123475 BC104270.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123475 BC104269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123475 NM_146461.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138464 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01273203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01991071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01991082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01476144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02142444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411701 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484767 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484766 - -
          Affymetrix Mouse Clariom S Array PSR0200033978.mm.1 NM_146461.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033978.mm.1 NM_146461.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033978.mm.1 TC0200004030.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6180347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772889 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772889 - -
          mouse-6 v1.1 (Illumina) scl19030.2.52_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19030.2.52_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2783920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2783920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline