Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732130_s1
          See other OLFR1204 GE Assays ›
          Gene Symbol:
          Olfr1204
          Gene Name:
          olfactory receptor 1204
          Gene Aliases:
          MOR232-6
          Chromosome Location:
          Chr.2: 88851952 - 88852881 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732130_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1204
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146463.2 NP_666674.2 1 162 142

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1204

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P209602 NM_146463.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P209602 NM_146463.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P209602 NM_146463.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P209602 NM_146463.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P209602 NM_146463.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P209602 NM_146463.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21411422 - -
          Whole Mouse Genome G4122A A_51_P209602 NM_146463.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P209602 NM_146463.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02110173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02110153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02110152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02110163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21411421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01329896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02047952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21411441 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473617 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473616 - -
          Affymetrix Mouse Clariom S Array PSR0200010121.mm.1 NM_146463.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010121.mm.1 NM_146463.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010121.mm.1 TC0200001333.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224002 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224002 - -
          mouse-6 v1.1 (Illumina) scl0258455.1_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258455.1_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224002 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline