Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732160_s1
          See other OLFR1254 GE Assays ›
          Gene Symbol:
          Olfr1254
          Gene Name:
          olfactory receptor 1254
          Gene Aliases:
          MOR231-13
          Chromosome Location:
          Chr.2: 89788406 - 89789350 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732160_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1254
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146476.1 NP_666687.1 1 159 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1254

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079741 NM_146476.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133663 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079741 NM_146476.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P482030 NM_146476.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133663 - -
          Whole Mouse Genome G4122A A_52_P482030 NM_146476.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131763 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01688853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01432100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02141109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01390913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484851 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484850 - -
          Affymetrix Mouse Clariom S Array PSR0200034106.mm.1 NM_146476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034106.mm.1 NM_146476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034106.mm.1 TC0200004072.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3870348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665975 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665975 - -
          mouse-6 v1.1 (Illumina) scl4270.1.1_206-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4270.1.1_206-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665975 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline