Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732180_s1
          See other OLFR890 GE Assays ›
          Gene Symbol:
          Olfr890
          Gene Name:
          olfactory receptor 890
          Gene Aliases:
          MOR162-3
          Chromosome Location:
          Chr.9: 38143137 - 38144081 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732180_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr890
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146481.2 NP_666692.2 1 457 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr890

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135178 NM_146481.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135178 NM_146481.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P388187 NM_146481.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433756 - -
          Whole Mouse Genome G4122A A_52_P388187 NM_146481.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07718903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08149150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08149145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07739398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08193521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08077815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584379 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584378 - -
          Affymetrix Mouse Clariom S Array PSR0900002778.mm.1 NM_146481.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002778.mm.1 NM_146481.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002778.mm.1 TC0900000387.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222814 - -
          mouse-6 v1.1 (Illumina) scl37103.1.16_84-S - -
          Illumina Sentrix mouse-6.v1 bead array 670372 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37103.1.16_84-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline