Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732191_s1
          See other OLFR889 GE Assays ›
          Gene Symbol:
          Olfr889
          Gene Name:
          olfactory receptor 889
          Gene Aliases:
          MOR162-2
          Chromosome Location:
          Chr.9: 38115798 - 38116727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732191_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr889
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146482.1 NP_666693.1 1 679 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr889

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1970895 NM_146482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1970895 NM_146482.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344896 NM_146482.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433654 - -
          Whole Mouse Genome G4122A A_51_P344896 NM_146482.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08058465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08058469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08058473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08058467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08058452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07858447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08058466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07858434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08058467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07858448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07469232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07858424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07858473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433698 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584377 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584376 - -
          Affymetrix Mouse Clariom S Array PSR0900002777.mm.1 NM_146482.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002777.mm.1 NM_146482.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002777.mm.1 TC0900000386.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6220193 - -
          Illumina mouse-6 v1.0 expression beadchip 106940114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641716 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213243 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641716 - -
          mouse-6 v1.1 (Illumina) GI_33239245-S - -
          mouse-6 v1.1 (Illumina) scl0258475.1_323-S - -
          Illumina Sentrix mouse-6.v1 bead array 106940114 - -
          Illumina Sentrix mouse-6.v1 bead array 6220193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258475.1_323-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106940114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641716 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641716 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline