Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732223_s1
          See other OLFR486 GE Assays ›
          Gene Symbol:
          Olfr486
          Gene Name:
          olfactory receptor 486
          Gene Aliases:
          MOR204-19
          Chromosome Location:
          Chr.7: 108171798 - 108172742 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732223_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr486
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146496.1 NP_666707.1 1 885 98

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr486

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094324 NM_146496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094324 NM_146496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P516972 NM_146496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P516972 NM_146496.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06389020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06389013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06389020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06142876 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566683 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566681 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566680 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566682 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500132 - -
          Illumina mouse-6 v1.0 expression beadchip 1500132 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625215 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625215 - -
          mouse-6 v1.1 (Illumina) scl0258489.1_109-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500132 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258489.1_109-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500132 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625215 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0