Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732296_s1
          See other OLFR944 GE Assays ›
          Gene Symbol:
          Olfr944
          Gene Name:
          olfactory receptor 944
          Gene Aliases:
          MOR171-19
          Chromosome Location:
          Chr.9: 39217359 - 39218294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732296_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr944
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146507.1 NP_666718.1 1 128 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr944

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518542 NM_146507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518542 NM_146507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518542 NM_146507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518542 NM_146507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518542 NM_146507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518542 NM_146507.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26437700 - -
          Whole Mouse Genome G4122A A_51_P518542 NM_146507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518542 NM_146507.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101649 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07616444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07616449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07616442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26437744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07616449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07850552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07616450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26437696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584489 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584488 - -
          Affymetrix Mouse Clariom S Array PSR0900002893.mm.1 NM_146507.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002893.mm.1 NM_146507.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002893.mm.1 TC0900000427.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101340450 - -
          Illumina mouse-6 v1.0 expression beadchip 780253 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710431 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710431 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242157 - -
          mouse-6 v1.1 (Illumina) GI_33239219-S - -
          mouse-6 v1.1 (Illumina) scl37080.1.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 101340450 - -
          Illumina Sentrix mouse-6.v1 bead array 780253 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37080.1.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780253 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710431 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710431 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline