Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732317_s1
          See other OLFR97 GE Assays ›
          Gene Symbol:
          Olfr97
          Gene Name:
          olfactory receptor 97
          Gene Aliases:
          MOR156-2
          Chromosome Location:
          Chr.17: 37231436 - 37232368 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732317_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146512.1 NP_666723.1 1 847 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr97

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183955 NM_146512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183955 BC134363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183955 NM_146512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183955 BC134363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P519620 NM_146512.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P519620 BC134363.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167992 - -
          Whole Mouse Genome G4122A A_52_P519620 NM_146512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P519620 BC134363.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13193136 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13193138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13571183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13193148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13571186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13193138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13383293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167984 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450867 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450868 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450866 - -
          Affymetrix Mouse Clariom S Array PSR1700019550.mm.1 NM_146512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019550.mm.1 NM_146512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019550.mm.1 TC1700002060.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5390020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258482 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258482 - -
          mouse-6 v1.1 (Illumina) scl49933.1.67_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49933.1.67_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2736190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2736190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline