Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732376_s1
          See other OLFR1186 GE Assays ›
          Gene Symbol:
          Olfr1186
          Gene Name:
          olfactory receptor 1186
          Gene Aliases:
          MOR230-5
          Chromosome Location:
          Chr.2: 88525501 - 88526590 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732376_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1186
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146530.2 NP_666741.2 1 865 139
          GenBank mRNA BC132093.1 - 1 40130370 139
          BC132095.1 - 1 40130372 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1186

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016134 BC132095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016134 NM_146530.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016134 BC132093.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112941 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016134 BC132095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016134 NM_146530.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016134 BC132093.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21410133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112941 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01554858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01554866 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01547270 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01554864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01314920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02269214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01829534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473599 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473598 - -
          Affymetrix Mouse Clariom S Array PSR0200010100.mm.1 NM_146530.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010100.mm.1 NM_146530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010100.mm.1 TC0200001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010101.mm.1 NM_146530.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010101.mm.1 NM_146530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010101.mm.1 TC0200001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010102.mm.1 NM_146530.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010102.mm.1 NM_146530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010102.mm.1 TC0200001322.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6370458 - -
          Illumina mouse-6 v1.0 expression beadchip 6370458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771929 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771929 - -
          mouse-6 v1.1 (Illumina) scl20664.1.1_232-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20664.1.1_232-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2936594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771929 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2936594 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0