Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732614_s1
          See other OLFR153 GE Assays ›
          Gene Symbol:
          Olfr153
          Gene Name:
          olfactory receptor 153
          Gene Aliases:
          GA_x6K02T2Q125-49033418-49034341, MOR177-5, Olfr4-2, V5
          Chromosome Location:
          Chr.2: 87532035 - 87532958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732614_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr153
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206823.1 NP_996559.1 1 341 138
          GenBank mRNA BC147769.1 - 1 580 9056708 138
          BC147777.1 - 1 580 9056711 138

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr153

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 BC147769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 BC147777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 BC147769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 BC147777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32248 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 BC147769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 BC147777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 NM_206823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 BC147769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 BC147777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 BC147769.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 BC147777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 BC147769.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 BC147777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406291 - -
          Whole Mouse Genome G4122A A_52_P304991 BC147769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304991 BC147777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304991 NM_206823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304991 BC147769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304991 BC147777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304991 NM_206823.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01261236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01261239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01672499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01564064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01261238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02213927 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02203269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01510890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01413240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01991283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01318893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01318892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01413239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01510893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01477038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01318890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01318898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01939684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02251717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01319869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02203268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473552 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473551 - -
          Affymetrix Mouse Clariom S Array PSR0200010064.mm.1 TC0200001296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010062.mm.1 TC0200001296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010063.mm.1 NM_206823.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010063.mm.1 NM_206823.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010063.mm.1 TC0200001296.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2630500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242718 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242718 - -
          mouse-6 v1.1 (Illumina) scl0110511.1_125-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0110511.1_125-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242718 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0