Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00732733_s1
          See other OLFR1462 GE Assays ›
          Gene Symbol:
          Olfr1462
          Gene Name:
          olfactory receptor 1462
          Gene Aliases:
          MOR202-13
          Chromosome Location:
          Chr.19: 13185734 - 13191592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00732733_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1462
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146693.1 NP_666904.1 1 345 140
          GenBank mRNA BC128026.1 - 1 349 40136583 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1462

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016445 NM_146693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016445 BC128026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016445 NM_146693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016445 BC128026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119255 NM_146693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119255 BC128026.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963865 - -
          Whole Mouse Genome G4122A A_51_P119255 NM_146693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119255 BC128026.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14191920 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14191915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14163073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963870 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461797 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461796 - -
          Affymetrix Mouse Clariom S Array PSR1900003031.mm.1 NM_146693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003031.mm.1 NM_146693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003031.mm.1 TC1900000267.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220008 - -
          Illumina mouse-6 v1.0 expression beadchip 5220008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710887 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710887 - -
          mouse-6 v1.1 (Illumina) scl6288.1.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6288.1.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710887 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline