Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00769480_m1
          See other KLRE1 GE Assays ›
          Gene Symbol:
          Klre1
          Gene Name:
          killer cell lectin-like receptor family E member 1
          Gene Aliases:
          Klre-1, NKG2I
          Chromosome Location:
          Chr.6: 129559032 - 129585827 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00769480_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klre1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153590.3 NP_705818.3 2 - 3 279 120
          GenBank mRNA AY100458.1 - 2 - 3 672 120
          BG145575.1 - 2 - 3 257 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001537.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klre1

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          natural killer cell activation
          positive regulation of interferon-gamma production
          positive regulation of natural killer cell mediated cytotoxicity
          transmembrane signaling receptor activity
          protein phosphatase binding
          protein homodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071716 AK041224.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071716 AY100458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071716 AF306663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071716 BX518942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071716 NM_153590.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47653 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140159 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071716 AK041224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071716 AY100458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071716 AF306663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071716 BX518942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071716 NM_153590.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844322 - -
          Whole Mouse Genome G4122A A_52_P592222 AY100458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592222 AF306663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592222 BG145575.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592222 NM_153590.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P383544 AK041224.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05853892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05499001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05853884 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05853890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05499000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05853889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05769777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05338846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05762560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05853890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05726224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05769778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05357353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542211 - -
          Affymetrix HT MG-430 PM Array Plate 1458642_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542205 - -
          Mouse Genome 430 2.0 Array 1458642_at - -
          Mouse Genome 430 2.0 Array 1458642_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243655_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243655_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600012531.mm.1 TC0600001537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012531.mm.1 NM_153590.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012531.mm.1 NM_153590.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012533.mm.1 TC0600001537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012533.mm.1 NM_153590.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012533.mm.1 NM_153590.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012532.mm.1 TC0600001537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012532.mm.1 NM_153590.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012532.mm.1 NM_153590.3 Refseq
          Mouse Expression Set 430 1458642_PM_at - -
          Mouse Expression Set 430 1458642_at - -
          Affymetrix HT Mouse Genome 430B Array 1458642_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458642_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631030 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239346 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239346 - -
          mouse-6 v1.1 (Illumina) scl29461.5.2_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29461.5.2_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239346 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0