Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00770943_m1
          See other POLR2D GE Assays ›
          Gene Symbol:
          Polr2d
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide D
          Gene Aliases:
          2310002G05Rik, 2610028L19Rik, HSRBP4, RBP4
          Chromosome Location:
          Chr.18: 31789159 - 31796701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00770943_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027002.3 NP_081278.1 2 - 3 264 95
          NM_027101.2 NP_081377.1 2 - 3 247 95
          GenBank mRNA AK009093.1 - 2 - 3 268 95
          AK011600.1 - 2 - 3 266 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000243.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Polr2d

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          mRNA export from nucleus in response to heat stress
          recruitment of 3'-end processing factors to RNA polymerase II holoenzyme complex
          cellular metabolic process
          positive regulation of translational initiation
          nucleotide binding
          single-stranded DNA binding
          single-stranded RNA binding
          catalytic activity
          DNA-directed RNA polymerase activity
          translation initiation factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800001737.mm.1 NM_027002.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001737.mm.1 NM_027002.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001737.mm.1 NM_027101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001737.mm.1 NM_027101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001737.mm.1 TC1800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001734.mm.1 NM_027002.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001734.mm.1 NM_027002.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001734.mm.1 NM_027101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001734.mm.1 NM_027101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001734.mm.1 TC1800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001735.mm.1 NM_027002.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001735.mm.1 NM_027002.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001735.mm.1 NM_027101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001735.mm.1 NM_027101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001735.mm.1 TC1800000243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001736.mm.1 NM_027002.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001736.mm.1 NM_027002.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001736.mm.1 NM_027101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001736.mm.1 NM_027101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001736.mm.1 TC1800000243.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline