Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00771682_g1
          See other S100A6 GE Assays ›
          Gene Symbol:
          S100a6
          Gene Name:
          S100 calcium binding protein A6 (calcyclin)
          Gene Aliases:
          2A9, 5B10, CALCYCLIN, Cacy, PRA
          Chromosome Location:
          Chr.3: 90612894 - 90614414 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00771682_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011313.2 NP_035443.1 2 - 3 497 70
          GenBank mRNA AF549497.1 - 2 - 3 205 70
          AK008241.1 - 2 - 3 208 70
          AK008941.1 - 2 - 3 208 70
          AK150408.1 - 2 - 3 208 70
          AK151144.1 - 2 - 3 208 70
          AK151587.1 - 2 - 3 208 70
          BC003832.1 - 2 - 3 260 3490662 70
          BC010774.1 - 2 - 3 222 4219642 70
          BM932500.1 - 2 - 3 509 70
          M37761.1 - 2 - 3 206 70
          X66449.1 - 299 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000809.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100a6

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          calcium ion binding
          protein binding
          tropomyosin binding
          zinc ion binding
          ion transmembrane transporter activity
          protein homodimerization activity
          S100 protein binding
          metal ion binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK008241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK008941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK150408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK151144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK151587.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK008241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK008941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK150408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK151144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281089 AK151587.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131269 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115386 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161910 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK008241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK008941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK150408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK151144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK151587.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK008241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK008941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK150408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK151144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281089 AK151587.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326137 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161910 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115386 - -
          Whole Mouse Genome G4122A A_51_P281089 AK008241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK008941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK150408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK151144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK151587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK008241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK008941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK150408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK151144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281089 AK151587.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118387 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161910 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03052293 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03052290 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02603145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02603129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02603127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02729495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02661616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03052290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02661611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02661615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02603129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03168786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02603130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02603119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02661613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02603128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326176 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493821 - -
          Affymetrix HT MG-430 PM Array Plate 1421375_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493820 - -
          Mouse Genome 430 2.0 Array 1421375_a_at - -
          Mouse Genome 430 2.0 Array 1421375_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20200_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20200_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006310.mm.1 NM_011313.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006310.mm.1 NM_011313.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006310.mm.1 TC0300000809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006311.mm.1 NM_011313.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006311.mm.1 NM_011313.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006311.mm.1 TC0300000809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006307.mm.1 NM_011313.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006307.mm.1 NM_011313.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006307.mm.1 TC0300000809.mm.1 MTA TC
          Mouse Expression Set 430 1421375_a_at - -
          Mouse Expression Set 430 1421375_PM_a_at - -
          Murine Genome U74v2 Set 92770_at - -
          Murine Genome U74v2 Set 92770_PM_at - -
          Mouse Genome 430A 2.0 Array 1421375_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1421375_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421375_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421375_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932445 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1690204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001690204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712120 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712120 - -
          mouse-6 v1.1 (Illumina) scl22942.3.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 1690204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22942.3.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712120 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712120 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22942.3.1_64-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline