Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00773197_gH
          See other OBOX5 GE Assays ›
          Gene Symbol:
          Obox5
          Gene Name:
          oocyte specific homeobox 5
          Gene Aliases:
          C87544
          Chromosome Location:
          Chr.7: 15750370 - 15759274 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00773197_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Obox5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145709.2 NP_663755.2 3 - 4 566 158
          GenBank mRNA BC052664.1 - 3 - 4 570 30053785 158
          DV653414.1 - 1 - 2 131 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Obox5

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 DV653414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 NM_145709.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 BC052664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 CJ052752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 DV653414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 NM_145709.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 BC052664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P178963 CJ052752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 DV653414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 NM_145709.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 BC052664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 CJ052752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 DV653414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 NM_145709.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 BC052664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P178963 CJ052752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 DV653414.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 NM_145709.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 BC052664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 CJ052752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 DV653414.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 NM_145709.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 BC052664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178963 CJ052752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 DV653414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 NM_145709.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 BC052664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 CJ052752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 DV653414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 NM_145709.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 BC052664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178963 CJ052752.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550187 - -
          Affymetrix HT MG-430 PM Array Plate 1436741_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550202 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550197 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550183 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550212 - -
          Mouse Genome 430 2.0 Array 1436741_PM_at - -
          Mouse Genome 430 2.0 Array 1436741_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252829_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 252829_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700001489.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001489.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001489.mm.1 NM_145709.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001482.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001482.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001482.mm.1 NM_145709.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001485.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001485.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001485.mm.1 NM_145709.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001483.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001483.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001483.mm.1 NM_145709.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001487.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001487.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001487.mm.1 NM_145709.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001486.mm.1 TC0700000212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001486.mm.1 NM_145709.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001486.mm.1 NM_145709.2 Refseq
          Mouse Expression Set 430 1436741_PM_at - -
          Mouse Expression Set 430 1436741_at - -
          Murine Genome U74v2 Set 130545_PM_at - -
          Murine Genome U74v2 Set 130545_at - -
          Affymetrix HT Mouse Genome 430B Array 1436741_at - -
          Affymetrix HT Mouse Genome 430B Array 1436741_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 390174 - -
          ABI Mouse Genome Survey Microarray 663861 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106620440 - -
          Illumina mouse-6 v1.0 expression beadchip 2940129 - -
          Illumina mouse-6 v1.0 expression beadchip 106620440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772986 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772986 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218255 - -
          mouse-6 v1.1 (Illumina) scl00252829.1_32-S - -
          mouse-6 v1.1 (Illumina) GI_21844548-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940129 - -
          Illumina Sentrix mouse-6.v1 bead array 106620440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00252829.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940129 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106620440 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218255 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline