Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00774453_g1
          See other CYPT3 GE Assays ›
          Gene Symbol:
          Cypt3
          Gene Name:
          cysteine-rich perinuclear theca 3
          Gene Aliases:
          1700006H02Rik, Ckt1r1
          Chromosome Location:
          Chr.X: 153558574 - 153559436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00774453_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cypt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173367.2 NP_775543.2 1 - 2 295 158
          GenBank mRNA AF463500.1 - 1 - 2 275 158
          AM040452.1 - 1 - 2 290 158
          BC048527.1 - 1 - 2 295 6772990 158

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001559.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cypt3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957533 NM_173367.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957533 AF463500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957533 AK005674.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101628 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957533 NM_173367.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957533 AF463500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957533 AK005674.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32753866 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101628 - -
          Whole Mouse Genome G4122A A_51_P212356 AK005674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212356 AM040452.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14553747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14553745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32753918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15029052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14526924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14674035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14646683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32753859 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10602721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602720 - -
          Affymetrix HT MG-430 PM Array Plate 1456192_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10602719 - -
          Mouse Genome 430 2.0 Array 1456192_x_at - -
          Mouse Genome 430 2.0 Array 1456192_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69361_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69361_at - -
          Affymetrix Mouse Clariom S Array PSR0X00009933.mm.1 NM_173367.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009933.mm.1 TC0X00001559.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009935.mm.1 NM_173367.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009935.mm.1 TC0X00001559.mm.1 MTA TC
          Mouse Expression Set 430 1456192_x_at - -
          Mouse Expression Set 430 1456192_PM_x_at - -
          Murine Genome U74v2 Set 165882_PM_r_at - -
          Murine Genome U74v2 Set 165882_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1456192_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1456192_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 594177 - -
          ABI Mouse Genome Survey Microarray 693085 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4280603 - -
          Illumina mouse-6 v1.0 expression beadchip 4280603 - -
          Illumina mouse-6 v1.0 expression beadchip 1980411 - -
          Illumina mouse-6 v1.0 expression beadchip 105360446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2664788 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253202 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253202 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223356 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2664788 - -
          mouse-6 v1.1 (Illumina) scl069361.2_47-S - -
          mouse-6 v1.1 (Illumina) scl0069361.1_139-S - -
          mouse-6 v1.1 (Illumina) GI_27734055-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980411 - -
          Illumina Sentrix mouse-6.v1 bead array 105360446 - -
          Illumina Sentrix mouse-6.v1 bead array 4280603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069361.1_139-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069361.2_47-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105360446 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2664788 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223356 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253202 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2664788 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline