Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00775056_m1
          See other C130079G13RIK GE Assays ›
          Gene Symbol:
          C130079G13Rik
          Gene Name:
          RIKEN cDNA C130079G13 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 59925214 - 59937949 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00775056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C130079G13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177661.3 NP_808329.1 5 - 6 971 134
          GenBank mRNA AK081819.1 - 5 - 6 972 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000479.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C130079G13Rik

          Panther Classification:

          Molecular Function -

          deacetylase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058606 AK140981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058606 NR_037189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955009 NM_177661.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955009 AK081819.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94581 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152033 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109623 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058606 AK140981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058606 NR_037189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955009 NM_177661.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955009 AK081819.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22175222 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109623 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152033 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04922809 - -
          Whole Mouse Genome G4122A A_51_P342418 NM_177661.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342418 AK081819.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118239 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109623 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152033 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02416730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02416710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02416714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02505082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22175202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02728031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02416717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02762459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02762449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02643149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02416718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02416714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02533986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03201153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02505083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22175238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10492290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492289 - -
          Affymetrix HT MG-430 PM Array Plate 1457645_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10492284 - -
          Mouse Genome 430 2.0 Array 1457645_PM_at - -
          Mouse Genome 430 2.0 Array 1457645_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229333_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229333_at - -
          Affymetrix Mouse Clariom S Array PSR0300003312.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003312.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003312.mm.1 TC0300000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003314.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003314.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003314.mm.1 TC0300000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003315.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003315.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003315.mm.1 TC0300000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003313.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003313.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003313.mm.1 TC0300000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003310.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003310.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003310.mm.1 TC0300000479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300003311.mm.1 NM_177661.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003311.mm.1 NM_177661.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300003311.mm.1 TC0300000479.mm.1 MTA TC
          Mouse Expression Set 430 1457645_at - -
          Mouse Expression Set 430 1457645_PM_at - -
          Murine Genome U74v2 Set 133984_at - -
          Murine Genome U74v2 Set 133984_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457645_at - -
          Affymetrix HT Mouse Genome 430B Array 1457645_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360253 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70047 - -
          Illumina mouse-6 v1.0 expression beadchip 70047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232462 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232462 - -
          mouse-6 v1.1 (Illumina) scl23151.6.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 70047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23151.6.1_70-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232462 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232462 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline