Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00775096_m1
          See other VPS37D GE Assays ›
          Gene Symbol:
          Vps37d
          Gene Name:
          vacuolar protein sorting 37D (yeast)
          Gene Aliases:
          Wbscr24
          Chromosome Location:
          Chr.5: 135072899 - 135078266 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00775096_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vps37d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177574.4 NP_808242.1 1 - 2 211 150
          GenBank mRNA AK090181.1 - 1 - 2 212 150
          AY352519.1 - 1 - 2 211 150

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vps37d

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114632 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114685 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114683 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114682 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114635 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114684 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064074 BC044789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064074 BU704550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064074 AK090181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064074 AK158287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064074 AY081953.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131974 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064074 BC044789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064074 BU704550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064074 AK090181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064074 AK158287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064074 AY081953.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24114640 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131974 - -
          Whole Mouse Genome G4122A A_52_P401629 BC044789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401629 BU704550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401629 AK090181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401629 AK158287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401629 AY081953.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129687 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04232176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04138021 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04232178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114682 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114685 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114683 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114684 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114635 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24114632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24114611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04232176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04788633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04199929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04309601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04972369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04496846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04981850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04981851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04972367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04284282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04386390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04364289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04822829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04972363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04973022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04690415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04232179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04123306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04232414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04668756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04990760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04232418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04695292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24114643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534425 - -
          Affymetrix HT MG-430 PM Array Plate 1457736_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534420 - -
          Mouse Genome 430 2.0 Array 1457736_PM_at - -
          Mouse Genome 430 2.0 Array 1457736_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194309_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194309_PM_at - -
          Mouse Expression Set 430 1457736_PM_at - -
          Mouse Expression Set 430 1457736_at - -
          Affymetrix HT Mouse Genome 430B Array 1457736_at - -
          Affymetrix HT Mouse Genome 430B Array 1457736_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311584 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3190142 - -
          Illumina mouse-6 v1.0 expression beadchip 3190142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234930 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234930 - -
          mouse-6 v1.1 (Illumina) scl25930.4_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25930.4_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234930 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234930 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25930.4_46-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline