Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00776138_gH
          See other CD1D2 GE Assays ›
          Gene Symbol:
          Cd1d2
          Gene Name:
          CD1d2 antigen
          Gene Aliases:
          CD1.2, Cd1b, Ly-38
          Chromosome Location:
          Chr.3: 86986586 - 86989533 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00776138_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd1d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289449.1 NP_001276378.1 1 - 2 121 120
          NM_007640.2 NP_031666.2 1 - 2 121 120
          NR_038188.1 1 - 2 121 120
          GenBank mRNA BC057149.1 - 1 - 2 121 5328694 120
          M63697.1 - 1 - 2 355 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000689.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd1d2

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          immune system process
          immune response
          antigen processing and presentation
          positive regulation of interleukin-2 biosynthetic process
          innate immune response
          positive regulation of interleukin-4 biosynthetic process
          antigen processing and presentation, exogenous lipid antigen via MHC class Ib
          positive regulation of NK T cell activation
          beta-2-microglobulin binding
          endogenous lipid antigen binding
          exogenous lipid antigen binding
          lipopeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971174 NM_007640.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971174 BC057149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971174 M63697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 NM_007640.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 BC057149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 M63697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 NM_007640.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 BC057149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P165705 M63697.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872075 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149056 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971174 NM_007640.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971174 BC057149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971174 M63697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 NM_007640.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 BC057149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 M63697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 NM_007640.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 BC057149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P165705 M63697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 NM_007640.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 BC057149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 M63697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 NM_007640.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 BC057149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165705 M63697.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22306003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22305988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22306009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22306008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04977333 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149056 - -
          Whole Mouse Genome G4122A A_52_P165705 NM_007640.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P165705 BC057149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P165705 M63697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P165705 NM_007640.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P165705 BC057149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P165705 M63697.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101213 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02336717 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02336736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02336723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22306032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22305997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02336736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02486882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03132359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02423119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02336731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22306032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22305994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02540001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10492959 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492961 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492958 - -
          Affymetrix HT MG-430 PM Array Plate 1421647_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10492957 - -
          Mouse Genome 430 2.0 Array 1421647_at - -
          Mouse Genome 430 2.0 Array 1421647_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300004560.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004560.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004560.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004557.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004557.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004557.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004562.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004562.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004562.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004556.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004556.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004556.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004558.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004558.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004558.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004559.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004559.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004559.mm.1 TC0300000689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004561.mm.1 NR_038188.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004561.mm.1 NR_038188.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004561.mm.1 TC0300000689.mm.1 MTA TC
          Mouse Expression Set 430 1421647_PM_at - -
          Mouse Expression Set 430 1421647_at - -
          Murine Genome U74v2 Set 101897_PM_g_at - -
          Murine Genome U74v2 Set 101896_PM_at - -
          Murine Genome U74v2 Set 101897_g_at - -
          Murine Genome U74v2 Set 101896_at - -
          Mouse Genome 430A 2.0 Array 1421647_at - -
          Mouse Genome 430A 2.0 Array 1421647_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421647_at - -
          Affymetrix HT Mouse Genome 430A Array 1421647_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 599068 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380739 - -
          Illumina mouse-6 v1.0 expression beadchip 6380739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257334 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257334 - -
          mouse-6 v1.1 (Illumina) scl23029.6.1_87-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23029.6.1_87-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257334 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline