Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00776306_mH
          See other KLRA6 GE Assays ›
          Gene Symbol:
          Klra6
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 6
          Gene Aliases:
          Ly49f, ly-49f
          Chromosome Location:
          Chr.6: 130013033 - 130026960 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00776306_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008464.2 NP_032490.1 4 - 5 497 110
          GenBank mRNA U10092.1 - 4 - 5 497 110

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra6

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004547 NM_008464.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004547 NM_008464.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P221993 U10092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P221993 NM_008464.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846234 - -
          Whole Mouse Genome G4122A A_52_P221993 U10092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221993 NM_008464.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05321431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05237089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05237120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05237108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548501 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548497 - -
          Affymetrix Murine Genome 6500A Array Msa13060_i_at - -
          Affymetrix Murine Genome 6500A Array Msa13060_r_i_at - -
          Affymetrix Murine Genome 6500A Array Msa13060_s_at - -
          Affymetrix Mouse Clariom S Array PSR0600025525.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025525.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025525.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025528.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025528.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025528.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025530.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025530.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025530.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025524.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025524.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025524.mm.1 NM_008464.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025529.mm.1 TC0600003253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025529.mm.1 NM_008464.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025529.mm.1 NM_008464.2 Refseq
          Murine Genome U74v2 Set 100674_PM_f_at - -
          Murine Genome U74v2 Set 100674_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360261 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 780181 - -
          Illumina mouse-6 v1.0 expression beadchip 100520184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673169 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229961 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673169 - -
          mouse-6 v1.1 (Illumina) scl093971.2_24-S - -
          mouse-6 v1.1 (Illumina) GI_6680591-S - -
          Illumina Sentrix mouse-6.v1 bead array 100520184 - -
          Illumina Sentrix mouse-6.v1 bead array 780181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl093971.2_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780181 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100520184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2673169 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2673169 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline