Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00777620_g1
          See other LAGE3 GE Assays ›
          Gene Symbol:
          Lage3
          Gene Name:
          L antigen family, member 3
          Gene Aliases:
          1110049G11Rik, Eso3, Itba2
          Chromosome Location:
          Chr.X: 74352162 - 74353618 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00777620_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lage3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025410.2 NP_079686.1 1 - 2 292 129
          GenBank mRNA AK002539.1 - 1 - 2 293 129
          AK004210.1 - 1 - 2 285 129
          AK134624.1 - 240 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002450.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lage3

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971328 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440413 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151564 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971329 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151564 - -
          Whole Mouse Genome G4122A A_52_P26826 AK004210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P26826 NM_025410.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P26826 AK002539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P26826 AK134624.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132394 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120909 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14700127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15137370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15140033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14682799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15140033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14832721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14808237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14832719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14682795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15047592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14682796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14747988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14747985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14703130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14747989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15087047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467668 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605318 - -
          Affymetrix HT MG-430 PM Array Plate 1449213_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605315 - -
          Mouse Genome 430 2.0 Array 1449213_PM_at - -
          Mouse Genome 430 2.0 Array 1449213_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66192_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66192_at - -
          Affymetrix Mouse Clariom S Array PSR0X00015574.mm.1 NM_025410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015574.mm.1 NM_025410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015574.mm.1 TC0X00002450.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015578.mm.1 NM_025410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015578.mm.1 NM_025410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015578.mm.1 TC0X00002450.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015580.mm.1 NM_025410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015580.mm.1 NM_025410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015580.mm.1 TC0X00002450.mm.1 MTA TC
          Mouse Expression Set 430 1449213_PM_at - -
          Mouse Expression Set 430 1449213_at - -
          Murine Genome U74v2 Set 104078_PM_g_at - -
          Murine Genome U74v2 Set 104077_PM_at - -
          Murine Genome U74v2 Set 104077_at - -
          Murine Genome U74v2 Set 161872_PM_f_at - -
          Murine Genome U74v2 Set 161872_f_at - -
          Murine Genome U74v2 Set 104078_g_at - -
          Murine Genome U74v2 Set 140870_PM_f_at - -
          Murine Genome U74v2 Set 140870_f_at - -
          Mouse Genome 430A 2.0 Array 1449213_at - -
          Mouse Genome 430A 2.0 Array 1449213_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449213_at - -
          Affymetrix HT Mouse Genome 430A Array 1449213_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893227 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090619 - -
          Illumina mouse-6 v1.0 expression beadchip 1090619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2679532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2679532 - -
          mouse-6 v1.1 (Illumina) scl066192.2_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066192.2_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2679532 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2679532 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066192.2_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline