Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00778060_sH
          See other POLR2H GE Assays ›
          Gene Symbol:
          Polr2h
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide H
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 20717826 - 20722272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00778060_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145632.2 NP_663607.1 2 243 124
          GenBank mRNA AK146429.1 - 2 268 124
          AK161617.1 - 2 264 124
          BC002306.1 - 2 243 3591968 124
          BC092087.1 - 2 153 5042770 124
          BC125612.1 - 2 227 40130127 124
          BC125638.1 - 2 227 40130153 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Polr2h

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          RNA polymerase I activity
          RNA polymerase II activity
          RNA polymerase III activity
          DNA binding
          DNA-directed RNA polymerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589175 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589176 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589177 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 AK146429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 AK161617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 NM_145632.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 BC092087.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122201 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 AK146429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 AK161617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 NM_145632.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 BC092087.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108731 BC002306.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122201 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03144671 - -
          Whole Mouse Genome G4122A A_51_P108731 BC002306.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108041 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122201 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12672584 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589177 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589176 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12672583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30589199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10400392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434524 - -
          Affymetrix HT MG-430 PM Array Plate 1424473_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400391 - -
          Affymetrix Mouse Gene 1.1 ST Array 10434523 - -
          Mouse Genome 430 2.0 Array 1424473_at - -
          Mouse Genome 430 2.0 Array 1424473_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 245841_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 245841_at - -
          Affymetrix Mouse Clariom S Array PSR1600002890.mm.1 NM_145632.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002890.mm.1 NM_145632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002890.mm.1 TC1600000291.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1424473_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424473_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 451990 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5890242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772936 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772936 - -
          mouse-6 v1.1 (Illumina) scl0245841.4_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0245841.4_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2874443 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874443 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772936 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0245841.4_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline