Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00779772_s1
          See other HIST1H2AB GE Assays ›
          Gene Symbol:
          Hist1h2ab
          Gene Name:
          histone cluster 1, H2ab
          Gene Aliases:
          H2A.1, H2a-53
          Chromosome Location:
          Chr.13: 23751088 - 23751592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00779772_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h2ab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175660.3 NP_783591.2 1 452 82

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h2ab

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883287 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P420466 NM_175660.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P420466 NM_175660.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P420466 NM_175660.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P420466 NM_175660.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883264 - -
          Whole Mouse Genome G4122A A_52_P420466 NM_175660.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P420466 NM_175660.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10703494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10703472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10703481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10703447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10703477 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11114053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883270 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404064 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404063 - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Mouse Expression Set 430 1438009_at - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1438009_PM_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Murine Genome U74v2 Set 94805_f_at - -
          Murine Genome U74v2 Set 94805_PM_f_at - -
          Murine Genome U74v2 Set 167053_PM_at - -
          Murine Genome U74v2 Set 167053_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1438009_PM_at - -
          Mouse Genome 430A 2.0 Array 1438009_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897729 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103120239 - -
          Illumina mouse-6 v1.0 expression beadchip 2680465 - -
          Illumina mouse-6 v1.0 expression beadchip 103120239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242304 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704873 - -
          mouse-6 v1.1 (Illumina) scl12329.1.1_313-S - -
          mouse-6 v1.1 (Illumina) GI_30061368-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680465 - -
          Illumina Sentrix mouse-6.v1 bead array 103120239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl12329.1.1_313-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103120239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2704873 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2704873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline