Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00779840_s1
          See other AIDA GE Assays ›
          Gene Symbol:
          Aida
          Gene Name:
          axin interactor, dorsalization associated
          Gene Aliases:
          2610208M17Rik, BC004835
          Chromosome Location:
          Chr.1: 183297060 - 183324501 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00779840_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aida
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181732.4 NP_859421.1 10 2839 123
          GenBank mRNA AK081494.1 - 10 2840 123
          AK149788.1 - 10 2590 123
          AK158103.1 - 10 2473 123
          AK172451.1 - 10 2532 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aida

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          dorsal/ventral pattern formation
          negative regulation of Wnt signaling pathway
          regulation of protein homodimerization activity
          negative regulation of JUN kinase activity
          negative regulation of JNK cascade
          determination of ventral identity
          negative regulation of determination of dorsal identity
          protein binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK172451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK158103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK149788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK081494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 NM_181732.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK172451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK158103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK149788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 AK081494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P462299 NM_181732.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC086763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 AK172451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC057183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC066829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 AK159318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC086763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 AK172451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC057183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 BC066829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P297332 AK159318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK172451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK158103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK149788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK081494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 NM_181732.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK172451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK158103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK149788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 AK081494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P462299 NM_181732.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC086763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 AK172451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC057183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC066829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 AK159318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC086763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 AK172451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC057183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 BC066829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P297332 AK159318.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09574650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09574655 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06408604 - -
          Whole Mouse Genome G4122A A_51_P462299 AK172451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK158103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK149788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK081494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 NM_181732.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK172451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK158103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK149788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 AK081494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P462299 NM_181732.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC086763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 AK172451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC057183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC066829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 AK159318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC086763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 AK172451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC057183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 BC066829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P297332 AK159318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P556247 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06408609 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09574655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25616140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25616139 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10384455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569894 - -
          Affymetrix HT MG-430 PM Array Plate 1460345_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423870_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10384454 - -
          Mouse Genome 430 2.0 Array 1423870_PM_at - -
          Mouse Genome 430 2.0 Array 1460345_at - -
          Mouse Genome 430 2.0 Array 1423870_at - -
          Mouse Genome 430 2.0 Array 1460345_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100014665.mm.1 NM_181732.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100014665.mm.1 NM_181732.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100014665.mm.1 TC0100001755.mm.1 MTA TC
          Mouse Expression Set 430 1423870_PM_at - -
          Mouse Expression Set 430 1423870_at - -
          Mouse Expression Set 430 1460345_PM_at - -
          Mouse Expression Set 430 1460345_at - -
          Mouse Genome 430A 2.0 Array 1460345_PM_at - -
          Mouse Genome 430A 2.0 Array 1423870_at - -
          Mouse Genome 430A 2.0 Array 1460345_at - -
          Mouse Genome 430A 2.0 Array 1423870_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423870_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460345_at - -
          Affymetrix HT Mouse Genome 430A Array 1460345_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423870_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 514836 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7040446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213265 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260459 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213265 - -
          mouse-6 v1.1 (Illumina) scl00108909.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00108909.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3104094 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260459 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0