Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00779901_s1
          See other SVS3B GE Assays ›
          Gene Symbol:
          Svs3b
          Gene Name:
          seminal vesicle secretory protein 3B
          Gene Aliases:
          9530004A22Rik
          Chromosome Location:
          Chr.2: 164254363 - 164256643 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00779901_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Svs3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173377.2 NP_775553.1 3 1097 150
          GenBank mRNA AK035235.1 - 3 1094 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Svs3b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114247 NM_173377.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114247 AK035235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114247 NM_173377.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114247 AK035235.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21817027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00549585 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114247 NM_173377.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114247 AK035235.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01222138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21816972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21817036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21816998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01219986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01219992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01801157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01801160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21816994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21816993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21817035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21816988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21816996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489457 - -
          Affymetrix Mouse Gene 1.1 ST Array 10489456 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329557_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329557_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200043572.mm.1 NM_173377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043572.mm.1 NM_173377.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043572.mm.1 TC0200005110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043574.mm.1 NM_173377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043574.mm.1 NM_173377.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043574.mm.1 TC0200005110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043573.mm.1 NM_173377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043573.mm.1 NM_173377.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043573.mm.1 TC0200005110.mm.1 MTA TC
          Mouse Expression Set 430 1417992_a_at - -
          Mouse Expression Set 430 1436782_PM_at - -
          Mouse Expression Set 430 1417993_at - -
          Mouse Expression Set 430 1436782_at - -
          Mouse Expression Set 430 1417992_PM_a_at - -
          Mouse Expression Set 430 1417993_PM_at - -
          Mouse Genome 430A 2.0 Array 1436782_PM_at - -
          Mouse Genome 430A 2.0 Array 1417993_at - -
          Mouse Genome 430A 2.0 Array 1436782_at - -
          Mouse Genome 430A 2.0 Array 1417992_a_at - -
          Mouse Genome 430A 2.0 Array 1417993_PM_at - -
          Mouse Genome 430A 2.0 Array 1417992_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 775621 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450086 - -
          Illumina mouse-6 v1.0 expression beadchip 6450086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258886 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258886 - -
          mouse-6 v1.1 (Illumina) scl18367.3.1_246-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18367.3.1_246-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258886 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258886 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0